Learn More
Bioworld Eriksson (ER) Medium, 25 L
Catalog No. 50254594
Encompass_Preferred
Eriksson (ER) MediumContains Macronutrients and Micronutrients as described by Eriksson (1965) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium Plant tissue culture tested
Catalog No. 50-254-594 Supplier Bioworld Supplier No. 306300233
Eriksson (ER) MediumContains Macronutrients and Micronutrients as described by Eriksson (1965) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium Plant tissue culture tested