Learn More
Bioworld Eriksson (ER) Medium w/ Vitamins, 25 L
Catalog No. 50254597
Encompass_Preferred
Eriksson (ER) MediumContains Macronutrients, Micronutrients, and Vitamins as described by Eriksson (1965) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-597 Supplier Bioworld Supplier No. 306300243
Eriksson (ER) MediumContains Macronutrients, Micronutrients, and Vitamins as described by Eriksson (1965) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested