Learn More
PHYTOTECHNOLOGY LABS ERIKSSON'S 1000X 100ML
Catalog No. NC1789699
Encompass_Preferred
Use a concentration of 1mL/L of medium to achieve proper final concentration.
Catalog No. NC1789699 Supplier PHYTOTECHNOLOGY LABS Supplier No. E330100ML
Use a concentration of 1mL/L of medium to achieve proper final concentration.