Learn More
Medchemexpress LLC Erinacine P | 291532-17-7 | 403923 | 492.60 | 1 MG
SDP
Catalog No. 5000415896
Encompass_Preferred
Erinacine P is a cyathane-type diterpenoid antibacterial agent. It is isolated from the biosynthesis of mycelia and submerged cultures of Hericium erinaceum. Erinacine P serves as a precursor for the production of important active components such as Erinacine C, A and B, and exhibits significant antibacterial activity against Aliivibrio fischeri.
  • Cyathane-type diterpenoid antibacterial agent.
  • Isolated from the biosynthesis of mycelia and submerged cultures of Hericium erinaceum.
  • Precursor for Erinacine C, A, and B.
  • Exhibits significant antibacterial activity against Aliivibrio fischeri.
Catalog No. 50-004-15896 Supplier Medchemexpress LLC Supplier No. HYN110191MG
Erinacine P is a cyathane-type diterpenoid antibacterial agent. It is isolated from the biosynthesis of mycelia and submerged cultures of Hericium erinaceum. Erinacine P serves as a precursor for the production of important active components such as Erinacine C, A and B, and exhibits significant antibacterial activity against Aliivibrio fischeri.
  • Cyathane-type diterpenoid antibacterial agent.
  • Isolated from the biosynthesis of mycelia and submerged cultures of Hericium erinaceum.
  • Precursor for Erinacine C, A, and B.
  • Exhibits significant antibacterial activity against Aliivibrio fischeri.