Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Eriochrome Black T

Catalog No. p-7023714
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1753622 100 g
AAA1753636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1753622 Supplier Thermo Scientific Chemicals Supplier No. A1753622
Only null left
Add to Cart

CAS: 1787-61-7 | C20H12N3NaO7S | 461.38 g/mol

Eriochrome black T is used as an indicator in analytical chemistry. It is also used in the detection of rare earth metals. It finds application in dyeing and printing of wool fabric, dyeing of silk and nylon as well as in leather dyeing. It can be used in the spectrophotometric determination of diphenhydramine hydrochloride in pharmaceutical preparations. As a complexometric indicator, it is useful in the determination of hardness of water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eriochrome black T is used as an indicator in analytical chemistry. It is also used in the detection of rare earth metals. It finds application in dyeing and printing of wool fabric, dyeing of silk and nylon as well as in leather dyeing. It can be used in the spectrophotometric determination of diphenhydramine hydrochloride in pharmaceutical preparations. As a complexometric indicator, it is useful in the determination of hardness of water.

Solubility
Soluble in water. Slightly soluble in alcohol and acetone.

Notes
Incompatible with strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1787-61-7
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) 461.38
MDL Number MFCD00003935
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
Synonym C.I. 14645; Mordant Black 11
PubChem CID 87355429
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

Beilstein 4121162
Merck Index 14,3667
Solubility Information Soluble in water. Slightly soluble in alcohol and acetone.
IUPAC Name (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium
Formula Weight 461.39
Quantity 100 g
Chemical Name or Material Eriochrome Black T
Hazard Category H319-H336-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 217-250-3
RTECSNumber QK2197000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.