Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol, Reagents

Catalog No. p-200022074
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-324 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-324 Supplier Reagents Holdings Llc Supplier No. CE255000100C
Only null left
Add to Cart
Add to Cart

Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol
Boiling Point 123°C
CAS 109-86-4, 1787-61-7
Synonym Water hardness indicator
Percent Purity 0.5%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.97
Quantity 100 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.