Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome Black T Indicator 1% w/w in Sodium Chloride (Water Hardness Indicator), Reagents

Catalog No. 01335394
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-394 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-394 Supplier Reagents Holdings Llc Supplier No. CE002400100A6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eriochrome Black T Indicator 1% w/w in Sodium Chloride (Water Hardness Indicator), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Eriochrome Black T Indicator 1% w/w in Sodium Chloride (Water Hardness Indicator)
CAS 7647-14-5, 1787-61-7
Boiling Point 1461°C
Packaging Poly Bottle
Specific Gravity 2.17
Synonym Water hardness indicator
Color Gray/Purple
pH 5 to 9
Quantity 100 mL
Physical Form Solid
Percent Purity 1%
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.