Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome™ Blue Black B, Solstice

Catalog No. 60046902 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-902 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-902 Supplier Solstice Supplier No. 4554025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicator (for complexometry)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3564-14-5
Molecular Formula C20H13N2NaO5S
Molecular Weight (g/mol) 416.38
MDL Number MFCD00003934
InChI Key YRFCCVCSEVISHI-GXTSIBQPSA-M
PubChem CID 23722702
IUPAC Name sodium;4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O

Specifications

Packaging Glass bottle
UN Number NONH for all modes of transport
Color Black
pH 9.5 (at 10 g/L 20°C)
Quantity 25 g
Physical Form Powder
Formula Weight 416.38g/mol
Chemical Name or Material Eriochrome™ Blue Black B
Hazard Statement P305 + P351 + P338
RTECSNumber 222-639-6
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.