Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome Blue Black R, Ricca Chemical
SDP

Catalog No. RDCE011525F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Pouch
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCE011525F 25 g Pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCE011525F Supplier Ricca Chemical Company Supplier No. RDCE011525F1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3564-14-5
Molecular Formula C20H13N2NaO5S
Molecular Weight (g/mol) 416.38
MDL Number MFCD00003934
InChI Key YRFCCVCSEVISHI-GXTSIBQPSA-M
PubChem CID 23722702
IUPAC Name sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-1-sulfonate
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging Pouch
Color Brown to Purple
Quantity 25 g
Physical Form Powder
Grade Indicator
Chemical Name or Material Eriochrome Blue Black R
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.