Learn More
TARGETMOL CHEMICALS INC ERIOCITRIN 10MG
SDP
Catalog No. 502576389
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Eriocitrin (Eriodictyol-7-O-Rutinoside) is powerful antioxidative flavonoid; (1) Prevents oxidative damages caused by acute exercise-induced oxidative stress.(2) Lipid-lowering effects in a rat model of high-fat diet. 2. Dietary Eriocitrin ameliorates diet-induced hepatic steatosis with activation of mitochondrial biogenesis. 3. Eriocitrin and Apigenin were identified as new potent inhibitors of human carbonic anhydrase VA isozyme. purity: 99%
Catalog No. 50-257-6389 Supplier TARGETMOL CHEMICALS INC Supplier No. T6S022110MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Eriocitrin (Eriodictyol-7-O-Rutinoside) is powerful antioxidative flavonoid; (1) Prevents oxidative damages caused by acute exercise-induced oxidative stress.(2) Lipid-lowering effects in a rat model of high-fat diet. 2. Dietary Eriocitrin ameliorates diet-induced hepatic steatosis with activation of mitochondrial biogenesis. 3. Eriocitrin and Apigenin were identified as new potent inhibitors of human carbonic anhydrase VA isozyme. purity: 99%