Learn More
Medchemexpress LLC HY-N0636 5mg Medchemexpress, Eriocitrin CAS:13463-28-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06365MG
Encompass_Preferred
Medchemexpress, HY-N0636 5mg Eriocitrin CAS:13463-28-0 Eriocitrin is a flavonoid isolated from lemon, which is a strong antioxidant agent. Eriocitrin could inhibit the proliferation of hepatocellular carcinoma cell lines by arresting cell cycle in S phase through up-regulation of p53, cyclin A, cyclin D3 and CDK6. Eriocitrin triggers apoptosis by activating mitochondria-involved intrinsic signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0299