Learn More
Medchemexpress LLC HY-N0637 5mg Medchemexpress, Eriodictyol CAS:552-58-9 Purity:>98%
SDP
Catalog No. 501933651
Encompass_Preferred
Medchemexpress, HY-N0637 5mg Eriodictyol CAS:552-58-9 Eriodictyol is a flavonoid isolated from the Chinese herb, with antioxidant and anti-inflammatory activity. Eriodictyol induces Nrf2 signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3651 Supplier Medchemexpress LLC Supplier No. HYN06375MG
Medchemexpress, HY-N0637 5mg Eriodictyol CAS:552-58-9 Eriodictyol is a flavonoid isolated from the Chinese herb, with antioxidant and anti-inflammatory activity. Eriodictyol induces Nrf2 signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.