Learn More
Cayman Chemical ErIodIctyol 7-O-glucosIde 10mg
SDP
Catalog No. 502912613
Encompass_Preferred
A flavanone glucoside; scavenges hydroxyl and superoxide anion radicals in cell-free assays (IC50s = 0.28 and 0.3 mM, respectively); activates Nrf2 in a reporter assay; protects against cisplatin-induced cytotoxicity in HRMCs, but not A549 or MDA-MB-231 cells, at 80 µM; attenuates neurological deficits and reduces infarct volume in a rat model of MCAO-induced cerebral ischemia at 30 mg/kg
Catalog No. 50-291-2613 Supplier Cayman Chemical Supplier No. 3600310 MG
A flavanone glucoside; scavenges hydroxyl and superoxide anion radicals in cell-free assays (IC50s = 0.28 and 0.3 mM, respectively); activates Nrf2 in a reporter assay; protects against cisplatin-induced cytotoxicity in HRMCs, but not A549 or MDA-MB-231 cells, at 80 µM; attenuates neurological deficits and reduces infarct volume in a rat model of MCAO-induced cerebral ischemia at 30 mg/kg