Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eriodictyol-7-O-glucoside | 38965-51-4 | 1 MG
SDP

Catalog No. 5000420213
Encompass_Preferred

Eriodictyol-7-O-glucoside is a flavonoid and a potent Nrf2 activator. It effectively scavenges hydroxyl radicals and superoxide anions and increases the nuclear localization of Nrf2, inducing the expression of Nrf2/ARE-dependent genes. This compound provides protection against oxygen and glucose deprivation (OGD)-induced oxidative insult and demonstrates neuroprotective efficacy in a rat model of focal cerebral ischemia. It is suitable for the study of stroke.

  • Purity: 99.42%
  • Molecular weight: 450.39
  • Formula: C21H22O11
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Structure classification: Flavonoids, flavanones, phenols, polyphenols
  • Increases Nrf2 protein level in a time-dependent and dose-dependent manner
  • Does not affect Nrf2 mRNA level but induces mRNA of HO-1, NQO1, and GCLC
  • Reduces oxygen and glucose deprivation (OGD)-induced cell death and intracellular ROS levels

Catalog No. 50-004-20213 Supplier Medchemexpress LLC Supplier No. HYN38471MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eriodictyol-7-O-glucoside is a flavonoid and a potent Nrf2 activator. It effectively scavenges hydroxyl radicals and superoxide anions and increases the nuclear localization of Nrf2, inducing the expression of Nrf2/ARE-dependent genes. This compound provides protection against oxygen and glucose deprivation (OGD)-induced oxidative insult and demonstrates neuroprotective efficacy in a rat model of focal cerebral ischemia. It is suitable for the study of stroke.

  • Purity: 99.42%
  • Molecular weight: 450.39
  • Formula: C21H22O11
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Structure classification: Flavonoids, flavanones, phenols, polyphenols
  • Increases Nrf2 protein level in a time-dependent and dose-dependent manner
  • Does not affect Nrf2 mRNA level but induces mRNA of HO-1, NQO1, and GCLC
  • Reduces oxygen and glucose deprivation (OGD)-induced cell death and intracellular ROS levels

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.