Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eriodictyol-7-O-glucoside | 38965-51-4 | MFCD18632528 | 99.8% | 450.39 g/mol | C21H22O11 | 10 MG
SDP

Catalog No. 5000289431
Encompass_Preferred

Eriodictyol-7-O-glucoside is a flavonoid glycoside used as a research reagent with reported antioxidant and Nrf2-activating activity in biological assays. Typical use includes in vitro biochemical and cell-based studies.

  • Flavonoid glycoside (eriodictyol-7-o-glucoside).
  • Molecular formula C21H22O11.
  • Molecular weight 450.39 g/mol.
  • CAS number 38965-51-4.
  • Purity 99.8% (manufacturer listed).
  • Solubility ≥ 2.5 mg/mL (5.55 mM); clear solution.
  • Supplied as 10 MG vial for laboratory use.

Catalog No. 50-002-89431 Supplier Medchemexpress LLC Supplier No. HYN384710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eriodictyol-7-O-glucoside is a flavonoid glycoside used as a research reagent with reported antioxidant and Nrf2-activating activity in biological assays. Typical use includes in vitro biochemical and cell-based studies.

  • Flavonoid glycoside (eriodictyol-7-o-glucoside).
  • Molecular formula C21H22O11.
  • Molecular weight 450.39 g/mol.
  • CAS number 38965-51-4.
  • Purity 99.8% (manufacturer listed).
  • Solubility ≥ 2.5 mg/mL (5.55 mM); clear solution.
  • Supplied as 10 MG vial for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.