Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erioglaucine Disodium Salt, Sigma-Aldrich

Catalog No. 01258954 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-954 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-954 Supplier MilliporeSigma Supelco Supplier No. 86114625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blue dye asuitable for diagnostic assay manufacturing, hematology, and histology, as well as for determining feeding behavior in Drosophila and the number of egg masses of Meloidogyne incognita

Erioglaucine disodium salt has been used as a coloring dye to determine the feeding behaviour in Drosophila. It has been used as a coloring dye for determining the number of egg masses of Meloidogyne incognita.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3844-45-9
Molecular Weight (g/mol) 792.85 g/mol

Specifications

Color Purple
Quantity 25 g
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.