Learn More
Santa Cruz Biotechnology ERK 1/2 Antibody (C-9), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC514302
Encompass
ERK 1/2 Antibody (C-9) is a high quality monoclonal ERK 1/2 antibody (also designated p44/42 MAPK antibody) suitable for the detection of the ERK 1/2 protein of mouse, rat and human origin. ERK 1/2 Antibody (C-9) is available as both the non-conjugated anti-ERK 1/2 antibody form, as well as multiple conjugated forms of anti-ERK 1/2 antibody, including agarose, HRP, PE, FITC and multiple Alexa Fluor conjugates.
Catalog No. NC1786844