Learn More
Cayman Chemical ERK InhIbItr 5mg
SDP
Catalog No. 502971326
Encompass_Preferred
A cell-permeable inhibitor that binds ERK2 near its docking domain (KD = 5 µM); blocks ERK-specific phosphorylation of Rsk-1 and TCF/Elk-1 but does not affect ERK1/2 phosphorylation by MEK1/2; completely prevents proliferation of several cancer cell lines
Catalog No. 50-297-1326 Supplier Cayman Chemical Supplier No. 159445 MG
A cell-permeable inhibitor that binds ERK2 near its docking domain (KD = 5 µM); blocks ERK-specific phosphorylation of Rsk-1 and TCF/Elk-1 but does not affect ERK1/2 phosphorylation by MEK1/2; completely prevents proliferation of several cancer cell lines