Learn More
Cayman Chemical ERK MAPK PhosphoThr202 Tyr20
SDP
Catalog No. 502886658
Encompass_Preferred
Antigen: phosphopeptide corresponding to amino acid residues surrounding the phospho-Thr202 and phospho-Tyr204 of rat ERK/MAPK • Host: rabbit • Cross Reactivity: (+) human and rat ERK/MAPK • Application(s): WB • ERK/MAPK is an integral component of cellular signaling during mitogenesis and differentiation of mitotic cells and also is thought to play a key role in learning and memory. The activity of this kinase is regulated by dual phosphorylation at Thr202 and Tyr204.
Catalog No. 50-288-6658 Supplier Cayman Chemical Supplier No. 100091791 EA
Antigen: phosphopeptide corresponding to amino acid residues surrounding the phospho-Thr202 and phospho-Tyr204 of rat ERK/MAPK • Host: rabbit • Cross Reactivity: (+) human and rat ERK/MAPK • Application(s): WB • ERK/MAPK is an integral component of cellular signaling during mitogenesis and differentiation of mitotic cells and also is thought to play a key role in learning and memory. The activity of this kinase is regulated by dual phosphorylation at Thr202 and Tyr204.