Learn More
TARGETMOL CHEMICALS INC ERK1/2 INHIBITOR 1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T112265MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ERK1/2 inhibitor 1 is a potent orally bioavailable ERK1/2 inhibitor showing 60% inhibition at 1 nM and an IC50 of 3.0 nM against ERK1 and ERK2 respectively. purity: 99%
Catalog No. 50-258-1550