Learn More
Medchemexpress LLC HY-112287 5mg Medchemexpress, ERK1/2 inhibitor 1 CAS:2095719-90-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1122875MG
Encompass_Preferred
Medchemexpress, HY-112287 5mg ERK1/2 inhibitor 1 CAS:2095719-90-5 ERK1/2 inhibitor 1 is a potent, orally bioavailable ERK1/2 inhibitor, showing 60% inhibition at 1 nM and an IC50 of 3.0 nM against ERK1 and ERK2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9722