Learn More
Medchemexpress LLC HY-126288 5mg Medchemexpress, ERK1/2 inhibitor 2 CAS:2095719-92-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1262885MG
Encompass_Preferred
Medchemexpress, HY-126288 5mg ERK1/2 inhibitor 2 CAS:2095719-92-7 ERK1/2 inhibitor 2 (Example 1) is a potent dual ERK1/2 inhibitor. ERK1/2 inhibitor 2 has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1095