Learn More
Medchemexpress LLC Erk1/2 inhibitor 2 (ASTX-029) | 2095719-92-7 | >98.0% | 584.04 g/mol | C29H31ClFN5O5 | 25MG
SDP
Catalog No. 5000236428
Encompass_Preferred
ASTX029 is a dual-mechanism extracellular signal-regulated kinase (ERK) 1/2 inhibitor developed as a clinical candidate. It inhibits ERK catalytic activity and ERK phosphorylation, with reported ERK2 IC50 ≈ 2.7 nM, and has demonstrated antineoplastic activity in preclinical models and early clinical evaluation. Supplied as a crystalline solid for research use, typical vendor-reported purity is ≥98% (CAS 2095719-92-7).
  • Potent dual-mechanism ERK inhibitor with low-nanomolar potency.
  • Inhibits both catalytic activity and phosphorylation of ERK.
  • Demonstrated antineoplastic activity in preclinical and early clinical studies.
  • Supplied as a crystalline solid suitable for in vitro and in vivo formulation.
  • Typical purity ≥98% by vendor testing.
  • CAS number 2095719-92-7 and molecular weight ≈584.04 g/mol.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-36428 Supplier Medchemexpress LLC Supplier No. HY12628825MG
ASTX029 is a dual-mechanism extracellular signal-regulated kinase (ERK) 1/2 inhibitor developed as a clinical candidate. It inhibits ERK catalytic activity and ERK phosphorylation, with reported ERK2 IC50 ≈ 2.7 nM, and has demonstrated antineoplastic activity in preclinical models and early clinical evaluation. Supplied as a crystalline solid for research use, typical vendor-reported purity is ≥98% (CAS 2095719-92-7).
  • Potent dual-mechanism ERK inhibitor with low-nanomolar potency.
  • Inhibits both catalytic activity and phosphorylation of ERK.
  • Demonstrated antineoplastic activity in preclinical and early clinical studies.
  • Supplied as a crystalline solid suitable for in vitro and in vivo formulation.
  • Typical purity ≥98% by vendor testing.
  • CAS number 2095719-92-7 and molecular weight ≈584.04 g/mol.
  • For research use only; not for human therapeutic use.