Learn More
Enzo Life Sciences ERK1/2 monoclonal antibody (12D4) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804061C100
Encompass_Preferred
Clone: 12D4. Isotype: IgG1. Immunogen: Peptide comprising the pThr-Glu-pTyr motif in activated ERK1 and ERK2 conjugated to KLH. Species reactivity: Human, Mouse, Rat, Dog. Specificity: Recognizes the pThr-Glu-pTyr motif of activated ERK1 and ERK2. Requires phosphorylation both at the threonine and the tyrosine site and does not cross-react with the non-phosphorylated form of the protein. Applications: ELISA, ICC, IHC, IP, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9765