Learn More
Enzo Life Sciences Erk 1/2 polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPMA001D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of rat Erk1. The sequence is completely conserved in hamster. UniProt: P21708. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Drosophila, Chicken, Bovine. Applications: Flow Cytometry, IHC (PS), IP, WB. Recommended dilutions: Flow Cytometry (10µg/ml). Immunohistochemistry (paraffin sections, 1:50). Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-666-525