Learn More
Enzo Life Sciences ERK1/2 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPMA001F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of rat Erk1. The sequence is completely conserved in hamster. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Drosophila, Chicken, Bovine. Applications: Flow Cytometry, IHC (PS), IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7278