Learn More
Abcam ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) In-Cell ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1264231X96TESTS
Encompass_Preferred
ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) In-Cell ELISA Kit is Cell-based (qualitative) ELISA for the measurement of ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) production in Mouse, Rat, Human in Adherent cells samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2741444