Learn More
ECM Bioscience ERK1(T207)/ERK2(T188)[conserved], phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience EP4101
Encompass_Preferred
The antibody detects 42 and 44 kDa* proteins corresponding to ERK1 (Thr-207) and ERK2 (Thr-188) on SDS-PAGE immunoblots of human A431 epithelial cells stimulated with calyculin A. It does not detect these ERK proteins in control cells or in blots treated with lambda phosphatase. The immunogen was Phospho-ERK2 (Thr-188) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding Thr-188 in mouse ERK2. This sequence is conserved in human, rat, chicken, and fish ERK2, and is highly conserved in ERK1 (Thr-207), ERK5 (Thr-224), and ERK7 (Thr-180). Accession #P63085. Applications = WB, E; Species Reactivity = Hu, Rt, Ms, Ck, F; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = ERK, p42, p44, MAPK T207, Thr-207, Thr207, T188, Thr-188, Thr188
Catalog No. 50-172-0781