Learn More
Carnabio Usa Inc ERK2(MAPK1) INACTIVE /50UG/FUL
Supplier:  Carnabio Usa Inc 041431050
Encompass
Erk2(MAPK1)[inactive], Full-length, Wild type, Amino Acid 1-360, NP_002736.3, Expressed in E. coli
Catalog No. 50-754-1035