Learn More
Sino Biological Recombinant Human ERK1/MAPK3 Protein (Full Length), Active
Catalog No. 502574333
Encompass_Preferred
Recombinant full length tag-free human ERK1 was expressed in E.coli cells and activated by active MEK1 in vitro.
Catalog No. 50-257-4333 Supplier Sino Biological Supplier No. M2910U50
Recombinant full length tag-free human ERK1 was expressed in E.coli cells and activated by active MEK1 in vitro.