Learn More
Apexbio Technology LLC ERK5-in-1, 10mg CAS# 1234479-76-5
SDP
Supplier:  Apexbio Technology LLC B838810
Encompass_Preferred
ERK5-IN-1 is a potent and selective ERK5 inhibitor. In vitro, ERK5-IN-1 effectively inhibits ERK5 biochemically with an IC50 of 162 nM. In cell experiments, ERK5-IN-1 has a cellular EC50 for inhibiting epidermal growth factor induced ERK5 autophosphorylation of 0.09 M. In cell-based analysis, it is at least 30 times more selective for ERK5 than LRRK2. It is also ERK5 selective for a group of other kinases. Available Oral Bioavailability. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8422