Learn More
Selleck Chemical LLC ERK5-IN-1 5mg 1435488-37-1 XMD17-109
SDP
Catalog No. 501364659
Encompass_Preferred
ERK5-IN-1 (XMD17-109) is a potent, and selective ERK5 inhibitor with IC50 of 162 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4659 Supplier Selleck Chemical LLC Supplier No. S73345MG
ERK5-IN-1 (XMD17-109) is a potent, and selective ERK5 inhibitor with IC50 of 162 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.