Learn More
Medchemexpress LLC HY-128341 5mg Medchemexpress, ERK5-IN-2 CAS:1888305-96-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1283415MG
Encompass_Preferred
Medchemexpress, HY-128341 5mg ERK5-IN-2 CAS:1888305-96-1 ERK5-IN-2 is an orally active, sub-micromolar, selective ERK5 inhibitor with IC50s of 0.82 μM, 3 μM for ERK5 and ERK5 MEF2D, respectively. ERK5-IN-2 does not interact with the BRD4 bromodomain. ERK5-IN-2 suppresses both tumor xenograft growth and basic fibroblast growth factor (bFGF) driven Matrigel plug angiogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9742