Learn More
Meso Scale Discovery ERKSTATCascade WCL
SDP
Catalog No. 5000078385
Encompass
The Meso Scale Discovery Phospho-ERK1/2 (Thr202/Tyr204) and Phospho-STAT3 (Tyr705) Whole Cell Lysate Kit is designed to measure endogenous levels of phosphorylated ERK1/2 and STAT3 in cell lysates The kit includes a 96-well plate coated with capture antibodies against total ERK1/2 and STAT3 It also includes detection antibodies against the phosphorylated forms of the proteins The kit provides a sensitive high-throughput method to monitor ERK and STAT3 pathway activation in cells It can be used for target identification/validation phenotypic screening and compound profiling studies The kit includes all necessary reagents and can process up to 80 samples in duplicate
Catalog No. 50-000-78385 Supplier Meso Scale Discovery Supplier No. K15116D2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Phospho-ERK1/2 (Thr202/Tyr204) and Phospho-STAT3 (Tyr705) Whole Cell Lysate Kit is designed to measure endogenous levels of phosphorylated ERK1/2 and STAT3 in cell lysates The kit includes a 96-well plate coated with capture antibodies against total ERK1/2 and STAT3 It also includes detection antibodies against the phosphorylated forms of the proteins The kit provides a sensitive high-throughput method to monitor ERK and STAT3 pathway activation in cells It can be used for target identification/validation phenotypic screening and compound profiling studies The kit includes all necessary reagents and can process up to 80 samples in duplicate