Learn More
Electron Microscopy Sciences ERL 4221 Cycloaliphatic Epoxide Resin 225 ML
SDP
Supplier:  Electron Microscopy Sciences 15005
Encompass_Preferred
Due to the supply shortage of ERL-4206 ERL-4221 has now been specially formulated to replace it. ERL 4206 is one of the main components of the low viscosity (Dr. Spur) embedding resin kit. ERL-4221 is a Cycloaliphatic Epoxide Resin.
Catalog No. 50-980-454