Learn More
Foxx Life Sciences Borosil® Narrow Mouth Erlenmeyer Conical Flasks with Interchangeable Glass Stopper 24/29, 100mL, 20/CS
Supplier:  Foxx Life Sciences 5020016
Encompass_Preferred
Borosil® brand Flasks are designed to deliver the best combination of thermal shock resistance along with physical strength & resistance to chemical attack. Borosil® Narrow Mouth Conical Erlenmeyer Flasks with Interchangeable Glass Stopper comes Uniform wall thickness distribution makes these flasks ideal for heating. The conical form makes suitable for mixing liquids. These Flasks can be closed with Glass Stopper. Avail. from 25mL to 2L. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-172-0531