Learn More
Foxx Life Sciences Borosil® Narroow Mouth Erlenmeyer Conical Flasks with Interchangeable Joint 24/29, 100mL, 10/CS
Supplier:  Foxx Life Sciences 5000016
Encompass_Preferred
Borosil® brand Flasks are designed to deliver the best combination of thermal shock resistance along with physical strength and resistance to chemical attack. Borosil® Narrow Mouth Conical Erlenmeyer Flasks with Interchangeable Joint comply with IS 1381. Uniform wall thickness distribution makes these flasks ideal for heating. The conical form makes suitable for mixing liquids. The easy-to-read scale and large labeling field helps easy marking. Avail. from 25mL to 500mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices.
Catalog No. 50-172-0395