Learn More
CELLTREAT Scientific Products 500mL Erlenmeyer Flask, Vent Cap, Plain Bottom, PC, Sterile
SDP
Catalog No. NC1964617
Encompass_Preferred
Designed for mammalian suspension culture applications as well as bacterial & yeast culture. Polycarbonate available with vent cap only. Vent caps with over-sized vent area feature 0.22μm hydrophobic membrane which allows for improved gas exchange without contamination. Avoid contamination with single-use, pre-sterilized flasks. Lightweight material is easier to handle than glass. Flasks include molded graduation lines. Individually bagged for easy storage and handling. Medical grade virgin Polycarbonate is autoclavable.
Catalog No. NC1964617 Supplier CELLTREAT Scientific Products Supplier No. 229829
Designed for mammalian suspension culture applications as well as bacterial & yeast culture. Polycarbonate available with vent cap only. Vent caps with over-sized vent area feature 0.22μm hydrophobic membrane which allows for improved gas exchange without contamination. Avoid contamination with single-use, pre-sterilized flasks. Lightweight material is easier to handle than glass. Flasks include molded graduation lines. Individually bagged for easy storage and handling. Medical grade virgin Polycarbonate is autoclavable.