Learn More
CELLTREAT Scientific Products CAP ONLY, 70mm, Erlenmeyer/Fernbach Vent Cap, Sterile
SDP
Catalog No. 50202102
Encompass_Preferred
CELLTREAT Scientific Products 70mm Erlenmeyer and Fernbach Flask CAP ONLY. Vent replacement Cap for Erlenmeyer and Fernbach Flasks. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 1/bag, 24/case.
Catalog No. 50-202-102 Supplier CELLTREAT Scientific Products Supplier No. 229895
CELLTREAT Scientific Products 70mm Erlenmeyer and Fernbach Flask CAP ONLY. Vent replacement Cap for Erlenmeyer and Fernbach Flasks. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 1/bag, 24/case.