Learn More
Chemglass Life Sciences Flask, Erlenmeyer, 250mL, PETG, with HDPE Vent Cap, Sterile
SDP
Catalog No. 501897582
Encompass
• PETG bottle with HDPE vent cap• Ideal for shaker culture applications.• Sterilized by E-Beam• Non-pyrogenic• Packaged in sterile bags• Breathable membrane• 0.22μm hydrophobic filter for gas exchange• Non-toxic and environmentally friendly
Catalog No. 50-189-7582 Supplier Chemglass Life Sciences Supplier No. CGE2092250
• PETG bottle with HDPE vent cap• Ideal for shaker culture applications.• Sterilized by E-Beam• Non-pyrogenic• Packaged in sterile bags• Breathable membrane• 0.22μm hydrophobic filter for gas exchange• Non-toxic and environmentally friendly