Learn More
Chemglass Life Sciences Flask, Erlenmeyer, 250mL, PETG, with HDPE Vent Cap, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN2092250
Encompass_Preferred
PETG flask with HDPE vent cap.Ideal for shaker culture applications.Sterilized by E-BeamNon-pyrogenic.Packaged in sterile bags.Breathable membrane.0. 22um hydrophobic filter for gas exchange.Non-toxic and environmentally friendly.
Catalog No. 50-189-7809