Learn More
Chemglass Life Sciences Flask, 2000mL, Erlenmeyer, Heavy Duty Rim, Neck Fits # 13 Rubber Stopper
SDP
Catalog No. NC0655722
Encompass_Preferred
A wide mouth flask especially recommended for use as a titration flask. For convenience, these flasks are graduated to show the approximate capacity, have an extra large marking spot, and heavy duty rims to reduce chipping. Flask, 2000mL, Erlenmeyer, Heavy Duty Rim, Neck Fits # 13 Rubber Stopper. *Stopper purchased seperately(
Catalog No. NC0655722 Supplier Chemglass Life Sciences Supplier No. CG85102L
A wide mouth flask especially recommended for use as a titration flask. For convenience, these flasks are graduated to show the approximate capacity, have an extra large marking spot, and heavy duty rims to reduce chipping. Flask, 2000mL, Erlenmeyer, Heavy Duty Rim, Neck Fits # 13 Rubber Stopper. *Stopper purchased seperately(