Learn More
Chemglass Life Sciences Flask, 500mL, Erlenmeyer, Heavy Duty Rim, Neck Fits # 10 Rubber Stopper
SDP
Catalog No. NC1732446
Encompass_Preferred
Flask, 500mL, Erlenmeyer, Heavy Duty Rim, Neck Fits # 10 Rubber Stopper. A wide mouth flask especially recommended for use as a titration flask. For convenience, these flasks are graduated to show the approximate capacity, have an extra large marking spot, and heavy duty rims to reduce chipping.
Catalog No. NC1732446 Supplier Chemglass Life Sciences Supplier No. CG8510500
Flask, 500mL, Erlenmeyer, Heavy Duty Rim, Neck Fits # 10 Rubber Stopper. A wide mouth flask especially recommended for use as a titration flask. For convenience, these flasks are graduated to show the approximate capacity, have an extra large marking spot, and heavy duty rims to reduce chipping.