Learn More
Chemglass Life Sciences Flask, Erlenmeyer, 500mL, PETG, with HDPE Vent Cap, Sterile
SDP
Supplier:  Chemglass Life Sciences CGE2092500
Encompass
• PETG bottle with HDPE vent cap• Ideal for shaker culture applications.• Sterilized by E-Beam• Non-pyrogenic• Packaged in sterile bags• Breathable membrane• 0.22μm hydrophobic filter for gas exchange• Non-toxic and environmentally friendly
Catalog No. 50-189-7583