Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Erlenmeyer Flask Clamp

Catalog No. 01922120 Shop All OHAUS Products
Change view
Click to view available options
For Use With (Equipment):
1 L Erlenmeyer Flask
2 L Erlenmeyer Flask
2.8 L Erlenmeyer Flask
3 L Erlenmeyer Flask
4 L Erlenmeyer Flask
5 L Erlenmeyer Flask
6 L Erlenmeyer Flask
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-922-120 6 L Erlenmeyer Flask
01-922-114 1 L Erlenmeyer Flask
01-922-115 2 L Erlenmeyer Flask
01-922-116 2.8 L Erlenmeyer Flask
01-922-117 3 L Erlenmeyer Flask
01-922-118 4 L Erlenmeyer Flask
01-922-119 5 L Erlenmeyer Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01-922-120 Supplier OHAUS™ Supplier No. 30400096
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OHAUS™ Erlenmeyer Flask Clamp is designed to securely grip and position various sizes of flasks.

  • Constructed of stainless steel
  • For use with flasks
  • Designed to securely hold flask in place
TRUSTED_SUSTAINABILITY

Specifications

Type Flask Clamp
Material Stainless Steel
For Use With (Equipment) 6 L Erlenmeyer Flask
For Use With (Application) Holding flasks
Description 6 Liter Erlenmeyer Flask Clamp
Color Silver
Jaw Type Four Interlocking
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.