Learn More
Hach Company Erlenmeyer flask PC 250 mL
Supplier: Hach Company 2089746
Flask-Erlenmeyer-Polycarbonate Capacity 250 mLGraduated (Molded-in graduations) flask features clarity toughness and strength Autoclavable however repeated autoclaving may result in loss of mechanical strength 6 Hollow stoppers (P/N 14480-06 pk/6) available separately 250 ml