Learn More
Foxx Life Sciences Borosil Narrow Mouth Glass Erlenmeyer Flasks, 50mL, 100/CS
Supplier:  Foxx Life Sciences 4980012
Encompass_Preferred
Borosil Erlenmeyer Flasks have uniform wall thickness, perfect for evenly distributed heat. The classic conical shape promotes effective mixing. Beaded rims confer durability and crack resistance. Graduations and marking spot further improve the functionality of these essential lab tools.Avail. in from 10mL to 5L. The 10mL option does not have graduations. Flasks comply with ISO/DIN 1773, with the exception of the 10mL, 125mL, and 150mL options. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-154-2845