Learn More
MTC Bio hybex™ Glass Erlenmeyer Flask with GL45 cap, 2000ml, each
Catalog No. NC1859333
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Dishes, plates and flasks, Flasks, Erlenmeyer flasks, hybex Glass Erlenmeyer Flasks with threaded GL45 cap, 2000ml. Made from high quality, borosilicate 3.3 glass, low coefficient of thermal expansion, high temperature resistance, permanent white enamel graduations and labeling area. Ideal for shaking samples and media preparation and storage. Each.
Catalog No. NC1859333 Supplier MTC Bio Supplier No. TB32202000
This item has a minimum qty of 2 per supplier requirements.
Dishes, plates and flasks, Flasks, Erlenmeyer flasks, hybex Glass Erlenmeyer Flasks with threaded GL45 cap, 2000ml. Made from high quality, borosilicate 3.3 glass, low coefficient of thermal expansion, high temperature resistance, permanent white enamel graduations and labeling area. Ideal for shaking samples and media preparation and storage. Each.