Learn More
MTC Bio hybex™ Glass Erlenmeyer Flask with GL45 cap, 500ml, 6/pk
Catalog No. NC1859331
Encompass_Preferred
Dishes, plates and flasks, Flasks, Erlenmeyer flasks, hybex Glass Erlenmeyer Flasks with threaded GL45 cap, 500ml. Made from high quality, borosilicate 3.3 glass, low coefficient of thermal expansion, high temperature resistance, permanent white enamel graduations and labeling area. Ideal for shaking samples and media preparation and storage. Pack of 6.
Catalog No. NC1859331 Supplier MTC Bio Supplier No. TB3220500
Dishes, plates and flasks, Flasks, Erlenmeyer flasks, hybex Glass Erlenmeyer Flasks with threaded GL45 cap, 500ml. Made from high quality, borosilicate 3.3 glass, low coefficient of thermal expansion, high temperature resistance, permanent white enamel graduations and labeling area. Ideal for shaking samples and media preparation and storage. Pack of 6.