Learn More
ABclonal Technology ERLIN1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A148431000UL
Encompass_Preferred
The protein encoded by this gene is part of a protein complex that mediates degradation of inositol 1,4,5-trisphosphate receptors in the endoplasmic reticulum. The encoded protein also binds cholesterol and regulates the SREBP signaling pathway, which promotes cellular cholesterol homeostasis. Defects in this gene have been associated with spastic paraplegia 62.
Catalog No. 50-001-08444